Anti-CD70 Goat Polyclonal Antibody

Supplier: Novus Biologicals
AF783
103223-124EA 649.32 USD
103223-124
Anti-CD70 Goat Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: CD70
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR