Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PTGIS Sheep Polyclonal Antibody

Supplier: Novus Biologicals
AF7788
Type: Primary Antibodies
Antigen: PTGIS
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 ug AF7788 103223-082 Each (100µG) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol PTGIS
Antigen Name Prostaglandin I2 Synthase
Reactivity Human
Clonality Polyclonal
Antibody Type Primary
Host Sheep
Isotype IgG
Antigen Synonyms CYP8A1, PGIS, PTGI, CYP8
Immunogen E. coli-derived recombinant human Prostaglandin I2 Synthase Arg21-Pro500 Accession # Q16647
Purification Immunogen affinity purified
Storage Buffer Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
Storage Temperature Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months, -20 to -70 °C as supplied. 1 month, 2 to 8 °C sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty