Anti-Complexin-1/2 Goat Polyclonal Antibody

Supplier: Novus Biologicals
AF7787-SP
103223-080EA 215.48 USD
103223-080
Anti-Complexin-1/2 Goat Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen:
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR