Anti-IGFBP6 Goat Polyclonal Antibody

Supplier: Novus Biologicals
AF776
103223-052EA 663.78 USD
103223-052
Anti-IGFBP6 Goat Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: IGFBP6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR