Anti-IL1R1 Goat Polyclonal Antibody

Supplier: Novus Biologicals
AF771
103222-992EA 707.17 USD
103222-992
Anti-IL1R1 Goat Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: IL1R1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR