Anti-PSMB10 Goat Polyclonal Antibody

Supplier: Novus Biologicals
AF7684
103222-970EA 663.78 USD
103222-970
Anti-PSMB10 Goat Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: PSMB10
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR