Anti-MKI67 Sheep Polyclonal Antibody

Supplier: Novus Biologicals
AF7649
103222-934EA 707.17 USD
103222-934
Anti-MKI67 Sheep Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: MKI67
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR