Anti-PSMA2 Sheep Polyclonal Antibody

Supplier: Novus Biologicals
AF7639
103222-912EA 663.78 USD
103222-912
Anti-PSMA2 Sheep Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: PSMA2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR