Anti-TH Sheep Polyclonal Antibody

Supplier: Novus Biologicals
AF7566
103222-894EA 678.25 USD
103222-894
Anti-TH Sheep Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: TH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR