Anti-NBL1 Goat Polyclonal Antibody

Supplier: Novus Biologicals
AF755-SP
103222-882EA 215.48 USD
103222-882
Anti-NBL1 Goat Polyclonal Antibody
Antibodies
Type: Primary Antibodies
Antigen: NBL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR