CVM Transport Media, for Chlamydia, Viruses, Mycoplasma, Hardy Diagnostics
R96 R96
10324-392PK 113.82 USD
CA10001-496 10324-392
CVM Transport Media, for Chlamydia, Viruses, Mycoplasma, Hardy Diagnostics
A multi-purpose transport for Chlamydia, Viruses, Ureaplasma, and Mycoplasma.
  • Transport difficult organisms
  • Specimens can be transported at refirgerated or room temperature for up to 48 hours
  • Break-resistant packaging
A multi-purpose transport for Chlamydia, Viruses, Ureaplasma, and Mycoplasma
The CVM has been extensively tested to assure viability for at least 48 hours for the following organisms: Herpes Simplex, Type I and II; Cytomegalovirus*; Influenza A and B; Respiratory Syncytial Virus; Echovirus; Adenovirus; Chlamydia trachomatis and pneumoniae; Mycoplasma hominis and pneumoniae; Ureaplasma urealyticum
The holding media is packaged in a tough, break-resistant, polypropylene centrifuge tube, with a secure screw-cap. Swabs not included. See R96KNP for the kit with swabs.
Difficult organisms are no problem for Hardy’s CVM Transport. Although the uninoculated CVM Transport is stored in the refrigerator, once the specimen is collected, it may be transported at refrigerator or room temperature for up to 48 hours.