Microslide: The Elements - Materials of the Universe
T-301
470200-590EA 11.45 USD
470200-590
Microslide: The Elements - Materials of the Universe
In this set students examine a few of the elements that can be found in their native, uncombined, form.
  • Displays common native elements
  • Text folder provides supplementary information
The accompanying text touches on atomic number and the periodic table and a simple chart depicts the process of atoms to rocks. An accompanying text folder provides the information about each microslide. Viewer available separately.
Set Includes: Carbon (Diamonds) (10X), Copper (5X), Gold (10X), Silver (15X), Sulphur (15X), Mercury (25X), Platinum (20X) and Lead (15X).