Ultra 6:1
40889
89400-774CS
505.82
USD
89400-774
Ultra 6:1
Ultra Etch
Wet chemical etching is an integral part of semiconductor manufacturing process. KMG’s line of ultra etchants (BOE w/surfactant) offers a variety of silicon dioxide etching rates and characteristics to meet your processing needs.
Specification Test Results
Assay (HF) | 7.13 - 7.33% |
Assay (NH4F) | 33.60 - 34.60% |
Color | 10 |
Appearance | Conform |
Surface Tension | 23 Dynes/cm |
Chloride | 2000 ppb |
Nitrate | 5000 ppb |
Phosphate | 400 ppb |
Sulfate | 1000 ppb |
Aluminum | 20 ppb |
Antimony | 5 ppb |
Arsenic | 5 ppb |
Barium | 10 ppb |
Boron | 10 ppb |
Cadmium | 10 ppb |
Calcium | 20 ppb |
Chromium | 10 ppb |
Cobalt | 10 ppb |
Copper | 10 ppb |
Gallium | 10 ppb |
Germanium | 10 ppb |
Gold | 10 ppb |
Iron | 20 ppb |
Lead | 10 ppb |
Lithium | 10 ppb |
Magnesium | 20 ppb |
Manganese | 10 ppb |
Nickel | 10 ppb |
Potassium | 20 ppb |
Silver | 10 ppb |
Sodium | 20 ppb |
Strontium | 10 ppb |
Tin | 20 ppb |
Titanium | 10 ppb |
Zinc | 10 ppb |
0.5µ Particle Count | 200 par/ml |
1.0µ Particle Count | 20 par/ml |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...