Ultra 15:1
40881
89400-768CS
505.82
USD
89400-768
Ultra 15:1
Ultra Etch
Wet chemical etching is an integral part of semiconductor manufacturing process. KMG’s line of ultra etchants (BOE w/surfactant) offers a variety of silicon dioxide etching rates and characteristics to meet your processing needs.
Specification Test Results
Assay (HF) | 3.07 - 3.27% |
Assay (NH4F) | 36.90 - 37.90% |
Appearance | Conform |
Surface Tension | 23 Dynes/cm |
Chloride | 2500 ppb |
Nitrate | 5000 ppb |
Phosphate | 500 ppb |
Sulfate | 1500 ppb% |
Aluminum | 20 ppb |
Antimony | 10 ppb |
Arsenic | 10 ppb |
Barium | 10 ppb |
Boron | 10 ppb |
Cadmium | 10 ppb |
Calcium | 20 ppb |
Chromium | 10 ppb |
Cobalt | 10 ppb |
Copper | 10 ppb |
Gallium | 10 ppb |
Germanium | 10 ppb |
Gold | 10 ppb |
Iron | 20 ppb |
Lead | 10 ppb |
Lithium | 10 ppb |
Magnesium | 20 ppb |
Manganese | 10 ppb |
Nickel | 10 ppb |
Potassium | 20 ppb |
Silver | 10 ppb |
Sodium | 50 ppb |
Strontium | 10 ppb |
Tin | 20 ppb |
Titanium | 10 ppb |
Zinc | 10 ppb |
0.5µ Particle Count | 200 par/ml |
1.0µ Particle Count | 20 par/ml |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...