ExoPure™ Immunoplates (Overall Exosome Isolation, Saliva, Fluorimetric Assay),BioVision Inc
M1015-100-B
75799-434EA 686.75 USD
75799-434
ExoPure™ Immunoplates (Overall Exosome Isolation, Saliva, Fluorimetric Assay),BioVision Inc
Immunoplates for Exosome capture and quantification from human biofluids (plasma, serum, urine, saliva) and from human or mouse cell media.
  • Various plates available depnding on downstream detection
  • Exosome capture
  • Quantification from cell media
Immunoplates for Exosome capture and quantification from human biofluids (plasma, serum, urine, saliva) and from human or mouse cell media. Transparent, white and black plates are available depending on the downstream detection approach (colorimetric, luminometric and fluorimetric respectively).