Buffered oxide etch 10:1
62060
89400-750CS
531.05
USD
89400-750
Buffered oxide etch 10:1
Buffered oxide etch
Buffered oxide etchants are used to etch thin films of silicon dioxide and shape contact and via openings.
Specification Test Results
Assay (HF) | 4.61 - 4.71% |
Assay (NH4F) | 35.70 - 36.70% |
Color |
|
Appearance | Conform |
Chloride | 2000 ppb |
Nitrate | 5000 ppb |
Phosphate | 500 ppb |
Sulfate | 1000 ppb |
Aluminum | 20 ppb |
Antimony | 5 ppb |
Arsenic | 5 ppb |
Barium | 10 ppb |
Boron | 10 ppb |
Cadmium | 10 ppb |
Calcium | 30 ppb |
Chromium | 10 ppb |
Cobalt | 10 ppb |
Copper | 10 ppb |
Gallium | 10 ppb |
Germanium | 10 ppb |
Gold | 10 ppb |
Iron | 50 ppb |
Lead | 20 ppb |
Lithium | 10 ppb |
Magnesium | 20 ppb |
Manganese | 10 ppb |
Nickel | 10 ppb |
Potassium | 25 ppb |
Silver | 10 ppb |
Sodium | 50 ppb |
Strontium | 10 ppb |
Tantalum | 10 ppb |
Thallium | 10 ppb |
Tin | 20 ppb |
Titanium | 20 ppb |
Zinc | 10 ppb |
0.5µ Particle Count | 160 par/ml |
1.0µ Particle Count | 8 par/ml |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...