Recombinant Human PPP1R14B, Abnova
Supplier: Abnova
H00026472-P01L
102925-306EA 350.77 USD
102925-306
Recombinant Human PPP1R14B, Abnova
Proteins and Peptides
Human PPP1R14B full-length ORF recombinant protein with GST-tag at N-terminal.
  • Preparation by in vitro wheat germ expression system
  • Glutathione sepharose 4 fast flow purification
  • 50 mM Tris-HCI, 10 mM reduced Glutathione, pH=8.0 in the elution buffer for storage
Used in Enzyme-linked immunoabsorbent assay, Western blot, Antibody production and Protein array.