Hydrochloric acid 37% GB for the electronics industry
Supplier: KMG
Synonyms:
Muriatic acid
64011
71009-358CS
980.17
USD
71009-358
Hydrochloric acid 37% GB for the electronics industry
Hydrochloric acid
Hydrochloric Acid is a key ingredient to remove surface metallic ions by forming soluble metal chlorides.
Formula:
HCl MW: 36.46 g/mol Boiling Pt: 110 °C (1013 hPa) Melting Pt: –30 °C Density: 1.18 g/cm³ (20 °C) |
MDL Number:
MFCD00011324 CAS Number: 7647-01-0 UN: 1789 ADR: 8,II |
Specification Test Results
Assay (HCl) | 37.00 - 38.00% |
Color | 10 APHA |
Appearance | Conform |
Phosphate | 50 ppb |
Sulfate | 200 ppb |
Sulfite | 700 ppb |
Aluminum | 5 ppb |
Antimony | 1 ppb |
Arsenic | 1 ppb |
Barium | 1 ppb |
Boron | 10 ppb |
Cadmium | 1 ppb |
Calcium | 10 ppb |
Chromium | 2 ppb |
Cobalt | 1 ppb |
Copper | 1 ppb |
Gallium | 1 ppb |
Germanium | 1 ppb |
Gold | 1 ppb |
Iron | 10 ppb |
Lead | 1 ppb |
Lithium | 1 ppb |
Magnesium | 1 ppb |
Manganese | 1 ppb |
Molybdenum | 1 ppb |
Nickel | 1 ppb |
Niobium | 1 ppb |
Potassium | 3 ppb |
Silver | 1 ppb |
Sodium | 3 ppb |
Strontium | 1 ppb |
Tin | 5 ppb |
Titanium | 1 ppb |
Vanadium | 1 ppb |
Zinc | 5 ppb |
0.50µ Particle Count | 50 par/ml |
1.00µ Particle Count | 10 par/ml |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...