HM Microtome Instrumentation Accessories, Thermo Scientific
Supplier: Thermo Scientific
630130 630130
16002-418EA 789.65 USD
16002-418 CA16002-418
HM Microtome Instrumentation Accessories, Thermo Scientific
Choose from multiple Thermo Scientific™ HM Microtome Instrumentation Accessories for use with Thermo Scientific™ HM Microtomes.
  • Automatic approach system
  • Direct sectioning possible after the automatic approach
  • Adaptable to HM 335 E, HM 340 E, HM 355 S, and HM 360 microtomes
  • Backlighting system
  • Illumination for block/knife relationship provided
  • Insertable into disposable blade carrier, knife carrier C, Standard, and S knife carrie
Ideal for semi-thin sectioning techniques. Useful for microscopic evaluation of the light reflex of the knife edge on the specimen.