Anti-CA9 Mouse Monoclonal Antibody [clone: GT14212]

Supplier: GeneTex
GTX629747
10167-472EA 577.14 USD
10167-472
Anti-CA9 Mouse Monoclonal Antibody [clone: GT14212]
Antibodies
Mouse Monoclonal antibody to Carbonic Anhydrase IX (carbonic anhydrase IX)

Type: Primary
Antigen: CA9
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR