Anti-PGAM1 Mouse Monoclonal Antibody [clone: GT256]

Supplier: GeneTex
GTX629745
10167-468EA 577.14 USD
10167-468
Anti-PGAM1 Mouse Monoclonal Antibody [clone: GT256]
Antibodies
Mouse Monoclonal antibody [GT256] to PGAM1 (phosphoglycerate mutase 1 (brain))

Type: Primary
Antigen: PGAM1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR