Anti-CDH1 Mouse Monoclonal Antibody [clone: GT477]

Supplier: GeneTex
GTX629691
10167-430EA 577.14 USD
10167-430
Anti-CDH1 Mouse Monoclonal Antibody [clone: GT477]
Antibodies
Mouse Monoclonal antibody [GT477] to E-Cadherin (cadherin 1, type 1, E-cadherin (epithelial))

Type: Primary
Antigen: CDH1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR