Anti-DDDDK tag Mouse Monoclonal Antibody [clone: GT1423]

Supplier: GeneTex
GTX629632
10167-426EA 577.14 USD
10167-426
Anti-DDDDK tag Mouse Monoclonal Antibody [clone: GT1423]
Antibodies
Mouse Monoclonal antibody [GT1423] to DDDDK tag

Type: Primary
Antigen: DDDDK tag
Clonality: Monoclonal
Clone: GT1423
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR