Anti-COL11A2 Mouse Monoclonal Antibody [clone: GT212]

Supplier: GeneTex
GTX629481
10167-408EA 655.44 USD
10167-408
Anti-COL11A2 Mouse Monoclonal Antibody [clone: GT212]
Antibodies
Mouse Monoclonal antibody [GT212] to COL11A2 (collagen, type XI, alpha 2)

Type: Primary
Antigen: COL11A2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR