Anti-CFDP1 Mouse Monoclonal Antibody [clone: GT9612]

Supplier: GeneTex
GTX629476
10167-398EA 577.14 USD
10167-398
Anti-CFDP1 Mouse Monoclonal Antibody [clone: GT9612]
Antibodies
Mouse Monoclonal antibody to CFDP1 (craniofacial development protein 1)

Type: Primary
Antigen: CFDP1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR