Anti-5-mC Mouse Monoclonal Antibody [clone: GT4111]

Supplier: GeneTex
GTX629448
10167-396EA 636.98 USD
10167-396
Anti-5-mC Mouse Monoclonal Antibody [clone: GT4111]
Antibodies
Mouse Monoclonal antibody [GT4111] to 5-Methylcytosine

Type: Primary
Antigen: 5-mC
Clonality: Monoclonal
Clone: GT4111
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR