Anti-X His tag Mouse Monoclonal Antibody [clone: GT359]

Supplier: GeneTex
GTX628914
89424-886EA 618.6 USD
89424-886
Anti-X His tag Mouse Monoclonal Antibody [clone: GT359]
Antibodies
Mouse Monoclonal antibody [GT359] to 6X His tag

Type: Primary
Antigen: His
Clonality: Monoclonal
Clone: GT359
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR