Anti-TET3 Mouse Monoclonal Antibody [clone: GT859]

Supplier: GeneTex
GTX628893
89424-862EA 727.5 USD
89424-862
Anti-TET3 Mouse Monoclonal Antibody [clone: GT859]
Antibodies
Mouse Monoclonal antibody [GT859] to TET3 (tet oncogene family member 3)

Type: Primary
Antigen: TET3
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR