Anti-SAMHD1 Mouse Monoclonal Antibody [clone: GT954]

Supplier: GeneTex
GTX628891
89424-858EA 582.76 USD
89424-858
Anti-SAMHD1 Mouse Monoclonal Antibody [clone: GT954]
Antibodies
Mouse Monoclonal antibody to SAMHD1 (SAM domain and HD domain 1)

Type: Primary
Antigen: SAMHD1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR