Anti-EGFR Mouse Monoclonal Antibody [clone: GT133]

Supplier: GeneTex
GTX628887
89424-850EA 582.76 USD
89424-850
Anti-EGFR Mouse Monoclonal Antibody [clone: GT133]
Antibodies
Mouse Monoclonal antibody [GT133] to EGFR (epidermal growth factor receptor (erythroblastic leukemia viral (v-erb-b) oncogene homolog, avian))

Type: Primary
Antigen: EGFR
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR