Anti-TAGLN Mouse Monoclonal Antibody [clone: GT336]

Supplier: GeneTex
GTX628831
89424-834EA 582.76 USD
89424-834
Anti-TAGLN Mouse Monoclonal Antibody [clone: GT336]
Antibodies
Mouse Monoclonal antibody to Transgelin (transgelin)

Type: Primary
Antigen: TAGLN
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR