Anti-TUBA1B Mouse Monoclonal Antibody [clone: GT114]

Supplier: GeneTex
GTX628802
89424-828EA 723.32 USD
89424-828
Anti-TUBA1B Mouse Monoclonal Antibody [clone: GT114]
Antibodies
Mouse Monoclonal antibody [GT114] to alpha Tubulin (tubulin, alpha 1b)

Type: Primary
Antigen: TUBA1B
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Drosophila, Rat, Zebra Fish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR