Anti-CD3 Mouse Monoclonal Antibody [clone: GT0013]

Supplier: GeneTex
GTX628462
89424-792EA 582.76 USD
89424-792
Anti-CD3 Mouse Monoclonal Antibody [clone: GT0013]
Antibodies
Mouse Monoclonal antibody [GT0013] to CD3

Type: Primary
Antigen:
Clonality: Monoclonal
Clone: GT0013
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR