Anti-Myc Mouse Monoclonal Antibody [clone: GT0002]

Supplier: GeneTex
GTX628259
89424-778EA 582.56 USD
89424-778
Anti-Myc Mouse Monoclonal Antibody [clone: GT0002]
Antibodies
Mouse Monoclonal antibody [GT0002] to Myc tag

Type: Primary
Antigen: Myc
Clonality: Monoclonal
Clone: GT0002
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR