Anti-SUZ12 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX60849
10162-758EA 491.76 USD
10162-758
Anti-SUZ12 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Suz12

Type: Primary
Antigen: SUZ12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR