Anti-RTF1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX60844
10162-748EA 574.63 USD
10162-748
Anti-RTF1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Rtf1

Type: Primary
Antigen: RTF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Drosophila
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR