Anti-L3MBTL1 Rabbit Polyclonal Antibody

GTX60834
10162-736EA 442.38 USD
10162-736
Anti-L3MBTL1 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to L3MBTL1

Type: Primary
Antigen: L3MBTL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR