Anti-HIST1H3A Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX60825
10162-718EA 589.99 USD
10162-718
Anti-HIST1H3A Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Histone H3R17me2K18ac (dimethyl Arg17, acetyl Lys18)

Type: Primary
Antigen: HIST1H3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR