Anti-ESR1 Mouse Monoclonal Antibody [clone: GT9077]

Supplier: GeneTex
GTX60809
10162-688EA 542.82 USD
10162-688
Anti-ESR1 Mouse Monoclonal Antibody [clone: GT9077]
Antibodies
Mouse monoclonal antibody [GT9077] to Estrogen Receptor alpha

Type: Primary
Antigen: ESR1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR