Anti-CDC7 Rabbit Polyclonal Antibody

GTX60805
10162-686EA 400.75 USD
10162-686
Anti-CDC7 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to Cdc73

Type: Primary
Antigen: CDC7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Drosophila
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR