Anti-SLC27A2 Mouse Monoclonal Antibody [clone: 6B3A9]

Supplier: GeneTex
GTX60795
10169-480EA 582.91 USD
10169-480
Anti-SLC27A2 Mouse Monoclonal Antibody [clone: 6B3A9]
Antibodies
Mouse Monoclonal Antibody [6B3A9] to SLC27A2

Type: Primary
Antigen: SLC27A2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR