Anti-CLGN Mouse Monoclonal Antibody [clone: 1C8B6]

Supplier: GeneTex
GTX60793
10169-476EA 582.91 USD
10169-476
Anti-CLGN Mouse Monoclonal Antibody [clone: 1C8B6]
Antibodies
Mouse Monoclonal Antibody [1C8B6] to CLGN

Type: Primary
Antigen: CLGN
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR