Anti-IL2RA Mouse Monoclonal Antibody [clone: 1B5D12]

Supplier: GeneTex
GTX60792
10169-474EA 613.46 USD
10169-474
Anti-IL2RA Mouse Monoclonal Antibody [clone: 1B5D12]
Antibodies
Mouse Monoclonal Antibody [1B5D12] to IL2RA

Type: Primary
Antigen: IL2RA
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Monkey, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR