Anti-ABCG5 Mouse Monoclonal Antibody [clone: 1B5E10]

Supplier: GeneTex
GTX60788
10169-466EA 582.91 USD
10169-466
Anti-ABCG5 Mouse Monoclonal Antibody [clone: 1B5E10]
Antibodies
Mouse Monoclonal Antibody [1B5E10] to ABCG5

Type: Primary
Antigen: ABCG5
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR