Anti-IL3RA Mouse Monoclonal Antibody [clone: 8E11C5]

Supplier: GeneTex
GTX60786
10169-462EA 613.46 USD
10169-462
Anti-IL3RA Mouse Monoclonal Antibody [clone: 8E11C5]
Antibodies
Mouse Monoclonal Antibody [8E11C5] to IL3RA

Type: Primary
Antigen: IL3RA
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR