Anti-MCAM Mouse Monoclonal Antibody [clone: 6C3E6]

Supplier: GeneTex
GTX60775
10169-440EA 617.81 USD
10169-440
Anti-MCAM Mouse Monoclonal Antibody [clone: 6C3E6]
Antibodies
Mouse Monoclonal Antibody [6C3E6] to MCAM

Type: Primary
Antigen: MCAM
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR