Anti-CD7 Mouse Monoclonal Antibody [clone: 4D4F8]

Supplier: GeneTex
GTX60756
10169-338EA 582.91 USD
10169-338
Anti-CD7 Mouse Monoclonal Antibody [clone: 4D4F8]
Antibodies
Mouse Monoclonal Antibody [4D4F8] to CD7

Type: Primary
Antigen: CD7
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR