Anti-KRT5 Mouse Monoclonal Antibody [clone: 2C2B4]

Supplier: GeneTex
GTX60749
10169-324EA 582.91 USD
10169-324
Anti-KRT5 Mouse Monoclonal Antibody [clone: 2C2B4]
Antibodies
Mouse Monoclonal antibody [2C2B4] to Cytokeratin 5

Type: Primary
Antigen: KRT5
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR