Anti-PIWIL4 Mouse Monoclonal Antibody [clone: 10G9B11]

Supplier: GeneTex
GTX60724
10169-354EA 442 USD
10169-354
Anti-PIWIL4 Mouse Monoclonal Antibody [clone: 10G9B11]
Mouse Monoclonal Antibody [10G9B11] to PIWIL4

Type: Primary
Antigen: PIWIL4
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR