Anti-C17orf53 Mouse Monoclonal Antibody [clone: 3E10H2]

Supplier: GeneTex
GTX60702
10168-736EA 442 USD
10168-736
Anti-C17orf53 Mouse Monoclonal Antibody [clone: 3E10H2]
Mouse Monoclonal Antibody [3E10H2] to C17ORF53

Type: Primary
Antigen: C17orf53
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR